AMBH324A5266 Ambeed, Inc.

(R)-4-Amino-3-(4-fluorophenyl)butanoic acid

  • MDL number MFCD11520152
  • Cas Number : 741217-33-4
  • Molecular Weight : 197.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/741217-33-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/741217-33-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart