AMBH324A5269 Ambeed, Inc.

(3aS,4S,6aR)-4-Methoxytetrahydrofuro[3,4-b]furan-2(3H)-one

  • MDL number MFCD10698799
  • Cas Number : 866594-60-7
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/866594-60-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/866594-60-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart