AMBH324A526C Ambeed, Inc.

(S)-1-((S)-1-(tert-Butoxycarbonyl)pyrrolidine-2-carbonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00057246
  • Cas Number : 15401-08-8
  • Molecular Weight : 312.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/15401-08-8.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/15401-08-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart