AMBH324A5271 Ambeed, Inc.

(S)-4-(tert-Butoxycarbonyl)-6,6-dimethylmorpholine-2-carboxylic acid

  • MDL number MFCD22690359
  • Cas Number : 1416444-82-0
  • Molecular Weight : 259.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1416444-82-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1416444-82-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart