AMBH324A5274 Ambeed, Inc.

((S)-1-((S)-1-Phenylethyl)pyrrolidin-3-yl)methanol

  • MDL number MFCD15526584
  • Cas Number : 173724-95-3
  • Molecular Weight : 205.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/173724-95-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/173724-95-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart