AMBH324A5279 Ambeed, Inc.

(R)-5-Bromo-3-[(1-methyl-2-pyrrolidinyl)methyl]-1H-indole

  • MDL number MFCD00912970
  • Cas Number : 143322-57-0
  • Molecular Weight : 293.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/143322-57-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/143322-57-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart