AMBH324A5280 Ambeed, Inc.

Z-Trp(Boc)-OH.DCHA

  • MDL number MFCD01318754
  • Cas Number : 218938-57-9
  • Molecular Weight : 619.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/218938-57-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/218938-57-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart