AMBH324A528B Ambeed, Inc.

(R)-tert-Butyl 2-ethylpyrrolidine-1-carboxylate

  • MDL number MFCD24466674
  • Cas Number : 876617-06-0
  • Molecular Weight : 199.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/876617-06-0.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/876617-06-0.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart