AMBH324A5299 Ambeed, Inc.

(S)-Methyl 6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-aminohexanoate hydrochloride

  • MDL number MFCD00153447
  • Cas Number : 201009-98-5
  • Molecular Weight : 418.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/201009-98-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/201009-98-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart