AMBH324A52A3 Ambeed, Inc.

(2R,3R,4R,5S)-6-(Ethylamino)hexane-1,2,3,4,5-pentaol

  • MDL number MFCD03789564
  • Cas Number : 14216-22-9
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/14216-22-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/14216-22-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart