AMBH324A52AB Ambeed, Inc.

2,2'-Anhydro-5-methyluridine

  • MDL number MFCD00233555
  • Cas Number : 22423-26-3
  • Molecular Weight : 240.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/22423-26-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/22423-26-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart