AMBH324A52BA Ambeed, Inc.

Boc-Tryptophanol

  • MDL number MFCD00235953
  • Cas Number : 82689-19-8
  • Molecular Weight : 290.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/82689-19-8.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/82689-19-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart