AMBH324A52D3 Ambeed, Inc.

(2R,4S)-Methyl 4-((tert-butoxycarbonyl)amino)pyrrolidine-2-carboxylate hydrochloride

  • MDL number MFCD11042659
  • Cas Number : 1279032-67-5
  • Molecular Weight : 280.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1279032-67-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1279032-67-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart