AMBH324A52D5 Ambeed, Inc.

Fmoc-L-beta-Lys(Boc)-OH

  • MDL number MFCD11227264
  • Cas Number : 219967-68-7
  • Molecular Weight : 468.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/219967-68-7.html

Safety Information

signalword Warning
pcodes P273
MSDS https://www.ambeed.com/products/219967-68-7.html
pictograms GHS07
hstatements H315-H413
Availability details for
Quantity
  • Availability
  • Add to Cart