AMBH324A52D9 Ambeed, Inc.

Uridine-5'-triphosphoric acid trisodium salt

  • MDL number MFCD00044310
  • Cas Number : 19817-92-6
  • Molecular Weight : 550.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/19817-92-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/19817-92-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart