AMBH324A52E2 Ambeed, Inc.

Dicyclohexylamine (S)-4-(((benzyloxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoate

  • MDL number MFCD00077017
  • Cas Number : 34897-61-5
  • Molecular Weight : 518.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/34897-61-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/34897-61-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart