AMBH324A52F4 Ambeed, Inc.

(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-difluorophenyl)propanoic acid

  • MDL number MFCD07371997
  • Cas Number : 1217820-78-4
  • Molecular Weight : 423.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1217820-78-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1217820-78-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart