AMBH324A5306 Ambeed, Inc.

(S)-2,5-Diaminopentanoic acid compound with (S)-2-aminosuccinic acid (1:1)

  • MDL number MFCD00058084
  • Cas Number : 3230-94-2
  • Molecular Weight : 265.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/3230-94-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/3230-94-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart