AMBH324A5314 Ambeed, Inc.

Potassium clavulanate

  • MDL number MFCD01710901
  • Cas Number : 61177-45-5
  • Molecular Weight : 237.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 50
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/61177-45-5.html

Safety Information

signalword Danger
pcodes P210-P261-P280-P305+P351+P338-P342+P311
MSDS https://www.ambeed.com/products/61177-45-5.html
pictograms GHS02,GHS08
hstatements H228-H317-H319-H334
Availability details for
Quantity
  • Availability
  • Add to Cart