AMBH324A532A Ambeed, Inc.

H-Lys(Z)-OtBu.HCl

  • MDL number MFCD00038898
  • Cas Number : 5978-22-3
  • Molecular Weight : 372.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/5978-22-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/5978-22-3.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart