AMBH324A5340 Ambeed, Inc.

Ammonium (Z)-2-(furan-2-yl)-2-(methoxyimino)acetate

  • MDL number MFCD03791040
  • Cas Number : 97148-39-5
  • Molecular Weight : 186.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/97148-39-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/97148-39-5.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart