AMBH324A535E Ambeed, Inc.

(R)-tert-Butyl (1-hydroxybut-3-en-2-yl)carbamate

  • MDL number MFCD19688626
  • Cas Number : 89985-86-4
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/89985-86-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/89985-86-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart