AMBH324A5362 Ambeed, Inc.

(R)-2-(2-Amino-3-(tritylthio)propanamido)acetic acid

  • Cas Number : 26988-61-4
  • Molecular Weight : 420.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/26988-61-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/26988-61-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart