AMBH324A5375 Ambeed, Inc.

(R)-1-(tert-Butoxycarbonyl)-3,3-dimethyl-1,3-azasilolidine-5-carboxylic acid

  • MDL number MFCD19689677
  • Cas Number : 268224-29-9
  • Molecular Weight : 259.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/268224-29-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/268224-29-9.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart