AMBH324A5377 Ambeed, Inc.

(S)-1-((S)-2-Aminopropanoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00037332
  • Cas Number : 13485-59-1
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/13485-59-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13485-59-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart