AMBH324A537C Ambeed, Inc.

(R)-4-(tert-Butoxy)-3-((tert-butoxycarbonyl)amino)-4-oxobutanoic acid

  • MDL number MFCD00236827
  • Cas Number : 77004-75-2
  • Molecular Weight : 289.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/77004-75-2.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/77004-75-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart