AMBH324A5382 Ambeed, Inc.

(2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-9-(((2R,3S)-3-Benzamido-2-hydroxy-3-phenylpropanoyl)oxy)-12-(benzoyloxy)-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-1H-7,11-methanocyclodeca[3,4]benzo[1,2-b]oxete-6,12b-diy

  • MDL number MFCD00869953
  • Cas Number : 33069-62-4
  • Molecular Weight : 853.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/33069-62-4.html

Safety Information

signalword Danger
pcodes P261-P280-P301+P310-P305+P351+P338-P342+P311
MSDS https://www.ambeed.com/products/33069-62-4.html
pictograms GHS05,GHS06,GHS08
hstatements H301-H315-H317-H318-H334
Availability details for
Quantity
  • Availability
  • Add to Cart