AMBH324A5393 Ambeed, Inc.

Ethyl 3-(dimethylamino)acrylate

  • MDL number MFCD00144269
  • Cas Number : 924-99-2
  • Molecular Weight : 143.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/924-99-2.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/924-99-2.html
pictograms GHS07
hstatements H315-H317-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart