AMBH324A53A2 Ambeed, Inc.

cis-Pyrrolidine-3,4-diol hydrochloride

  • MDL number MFCD21602660
  • Cas Number : 186393-21-5
  • Molecular Weight : 139.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/186393-21-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/186393-21-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart