AMBH324A53AC Ambeed, Inc.

(S)-tert-Butyl 3-(2-ethoxy-2-oxoethyl)morpholine-4-carboxylate

  • MDL number MFCD23701548
  • Cas Number : 761460-04-2
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/761460-04-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/761460-04-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart