AMBH324A53B0 Ambeed, Inc.

(2S,3R)-2-(((Benzyloxy)carbonyl)amino)-3-(tert-butoxy)butanoic acid

  • MDL number N/A
  • Cas Number : 16966-09-9
  • Molecular Weight : 309.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/16966-09-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/16966-09-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart