AMBH324A53C2 Ambeed, Inc.

H-Tyr(tBu)-OMe.HCl

  • MDL number MFCD00153466
  • Cas Number : 51482-39-4
  • Molecular Weight : 287.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/51482-39-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/51482-39-4.html
pictograms GHS07
hstatements H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart