AMBH324A53D3 Ambeed, Inc.

Dicyclohexylamine (S)-2-((tert-butoxycarbonyl)amino)-3-(furan-2-yl)propanoate

  • MDL number MFCD01860894
  • Cas Number : 331730-08-6
  • Molecular Weight : 436.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/331730-08-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/331730-08-6.html
pictograms GHS07
hstatements H302-H315-H319-H335-H413
Availability details for
Quantity
  • Availability
  • Add to Cart