AMBH324A53E0 Ambeed, Inc.

(1S)-(-)-Camphanic acid

  • MDL number MFCD00044948
  • Cas Number : 13429-83-9
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/13429-83-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/13429-83-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart