AMBH324A53E8 Ambeed, Inc.

N6-Benzoyladenosine

  • MDL number MFCD00056988
  • Cas Number : 4546-55-8
  • Molecular Weight : 371.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/4546-55-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/4546-55-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart