AMBH324A53EC Ambeed, Inc.

(S)-(9H-Fluoren-9-yl)methyl (1-oxopropan-2-yl)carbamate

  • MDL number MFCD01318541
  • Cas Number : 146803-41-0
  • Molecular Weight : 295.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/146803-41-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/146803-41-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart