AMBH324A53F2 Ambeed, Inc.

tert-Butyl ((1R,2R)-2-hydroxycyclopentyl)carbamate

  • MDL number MFCD11656037
  • Cas Number : 454170-16-2
  • Molecular Weight : 201.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/454170-16-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/454170-16-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart