AMBH324A53F9 Ambeed, Inc.

Sodium 1,4-diethoxy-1,4-dioxobut-2-en-2-olate

  • MDL number MFCD00035571
  • Cas Number : 40876-98-0
  • Molecular Weight : 210.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/40876-98-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/40876-98-0.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart