AMBH324A53FA Ambeed, Inc.

Fmoc-Asp(EDANS)-OH

  • MDL number MFCD02094140
  • Cas Number : 182253-73-2
  • Molecular Weight : 603.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/182253-73-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/182253-73-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart