AMBH324A53FC Ambeed, Inc.

Valganciclovir hydrochloride

  • MDL number MFCD08460118
  • Cas Number : 175865-59-5
  • Molecular Weight : 390.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/175865-59-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/175865-59-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart