AMBH324A540C Ambeed, Inc.

Boc-D-Cys(pMeBzl)-OH

  • MDL number MFCD00076924
  • Cas Number : 61925-78-8
  • Molecular Weight : 325.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/61925-78-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/61925-78-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart