AMBH324A5422 Ambeed, Inc.

D-(+)-Maltose monohydrate

  • MDL number MFCD00149343
  • Cas Number : 6363-53-7
  • Molecular Weight : 360.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/6363-53-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/6363-53-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart