AMBH324A5425 Ambeed, Inc.

4-(trans-4-Pentylcyclohexyl)benzoic acid

  • MDL number MFCD06208360
  • Cas Number : 65355-30-8
  • Molecular Weight : 274.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/65355-30-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/65355-30-8.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart