AMBH324A5428 Ambeed, Inc.

(E)-4-methoxy-4-oxobut-2-enoic acid(98%)

  • MDL number MFCD00063174
  • Cas Number : 2756-87-8
  • Molecular Weight : 130.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2756-87-8.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/2756-87-8.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart