AMBH324A542A Ambeed, Inc.

(S)-Methyl 3-(1-amino-2-hydroxyethyl)benzoate hydrochloride

  • MDL number MFCD12910849
  • Cas Number : 1391447-69-0
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1391447-69-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1391447-69-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart