AMBH324A542C Ambeed, Inc.

3-Cyclopentylacrylonitrile

  • MDL number MFCD11857755
  • Cas Number : 591769-05-0
  • Molecular Weight : 121.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/591769-05-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/591769-05-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart