AMBH324A5433 Ambeed, Inc.

(3aS,4S,6S,7aR)-2-Isobutyl-3a,5,5-trimethylhexahydro-4,6-methanobenzo[d][1,3,2]dioxaborole

  • MDL number MFCD09837626
  • Cas Number : 84110-34-9
  • Molecular Weight : 236.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/84110-34-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/84110-34-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart