AMBH324A5437 Ambeed, Inc.

(S)-6,7-Difluorochroman-4-amine

  • MDL number MFCD07374041
  • Cas Number : 1213126-25-0
  • Molecular Weight : 185.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1213126-25-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1213126-25-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart