AMBH324A5438 Ambeed, Inc.

Z-Gln(Trt)-OH

  • MDL number MFCD00144845
  • Cas Number : 132388-60-4
  • Molecular Weight : 522.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/132388-60-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/132388-60-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart