AMBH324A543E Ambeed, Inc.

(trans,trans)-3,4,5-Trifluorophenyl 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate

  • MDL number MFCD11053486
  • Cas Number : 132123-45-6
  • Molecular Weight : 382.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/132123-45-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/132123-45-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart